
2-Fluoro-5-nitrobenzaldehyde 97%
Catalog # ACRO371730010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Beige to light brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:56 to 60 °C
- GC:≥96.0 %
Specification Test Results
About this item
:
:169.11 g/mol
Melting Pt:57…60 °C
:GHS07
Storage Temperature:Ambient
CAS Number:27996-87-8
Formula:C₇H₄FNO₃
MDL Number:MFCD00042298