
(1S,2R,3S,4S)-2,3-Dihydroxy-4-(hydroxymethyl)-1-aminocyclopentane hydrochloride 95%, 98% ee
ACRO362530010
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥94 %
- Enantiomeric excess:≥97.5 %
- Specific optical rotation:+25° to +35° (20°C, 589 nm) (c=1 in water)
:
:183.64 g/mol
:124…124.6 °C
:GHS07
:Ambient
:220497-88-1
:C₆H₁₄ClNO₃
:MFCD06799069