Order Entry
United Kingdom
ContactUsLinkComponent
β-Methyl-DL-phenylalanine hydrochloride 99%
β-Methyl-DL-phenylalanine hydrochloride 99%
Catalog # ACRO202660010
CAS Number:  
undefined
β-Methyl-DL-phenylalanine hydrochloride 99%
Catalog # ACRO202660010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Titration with HClO4:
    ≥98.5 %

Specification Test Results

About this item

Synonyms: β-Methyl-3-phenyl-DL-alanine HCl, 2-Amino-3-phenylbutanoic acid hydrochloride, (±)-β-Methylphenylalanine hydrochloride
 :
 :215.68 g/mol
Melting Pt:210 °C (dec.)
Storage Temperature:Ambient
CAS Number:80997-87-1
Formula:C₁₀H₁₄NO₂Cl
MDL Number:MFCD00012623