Order Entry
United Kingdom
ContactUsLinkComponent
3-Methoxy-2-nitrobenzaldehyde 97%
Irritant
3-Methoxy-2-nitrobenzaldehyde 97%
Certificates
undefined
Irritant
3-Methoxy-2-nitrobenzaldehyde 97%

Specification Test Results

  • Appearance:
    White to yellow to beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    97 to 103 °C
  • GC:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together