Elaidic acid 98%
: (E)-Octadec-9-enoic acid
:112-79-8
:
:C₁₈H₃₄O₂
:MFCD00063954
:288 °C (100 mmHg)
:282.47 g/mol
:43…45 °C
:>110 °C
:GHS07
:Freezer
- Environmentally Preferable:
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:42 to 44 °C
- GC:≥97.5 %
- Chloride (Cl):≤0.5 ppm
- Sulfate (SO4):≤1 ppm
- Arsenic (As):≤0.01 ppm
- Iron (Fe):≤0.2 ppm
- Heavy metals (as Pb):≤0.2 ppm
Frequently Bought Together