Packers, wide neck, amber glass
Catalog # EPSC323-32A
Supplier: EP SCIENTIFIC
Specifications
- Closure type:Solid cap
- Capacity:1000 ml
- Description:Solid PP caps with 0,015" thick PTFE liners, 53-400
- Pk:12
Specifications
About this item
Jars are preferred for solids, semi-solids and hazardous waste samples.
- It protects the light-sensitive samples
- Recommended for semivolatiles, pesticides and metals sampling