Aluminium bromide, anhydrous 98+%, extra pure
: Aluminum bromide
:7727-15-3
:
:AlBr₃
:MFCD00003421
:266.69 g/mol
:265 °C (760 mmHg)
:97 °C
:3,2 g/cm³ (20 °C)
:Refrigerator
:1725
:8,II
:GHS07
:GHS05
- Environmentally Preferable:
- Appearance:White to yellow to pink to grey Crystalline powder or chunks
- Titration Complexometric:≥98.0 %
- Chloride (Cl):≤2000 ppm
- Sulfate (SO4):≤200 ppm
- Iron (Fe):≤50 ppm
- Lead (Pb):≤50 ppm
Frequently Bought Together