
4-Chloro-5-nitrobenzene-1,2-diamine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:67073-39-6
:
Formula:C₆H₆ClN₃O₂
MDL Number:MFCD03427594
:187.583 g/mol
Melting Pt:189…193 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Red Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together