


Dimethyltitanocene 5% (w/w) in toluene, AcroSeal®
301761000.
:
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
- Appearance:Orange to dark red Solution
- NMR (1H-NMR):Consistent with structure
- Strength:5 % (approximately)
:
:208.13 g/mol
:0.9 g/cm³
:Freezer
:1294
:3,II
:GHS07
:GHS08
:GHS02
:1271-66-5
:C₁₂H₁₆Ti
:MFCD02093882