Order Entry
United Kingdom
ContactUsLinkComponent
 
 
ɑ-Cyclopentyl-DL-mandelic acid 98%
  ACRO250970050
 :  
undefined
 
ɑ-Cyclopentyl-DL-mandelic acid 98%
  ACRO250970050
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    White to beige Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    144 to 150 °C
  • Titration with NaOH:
    ≥97.5 %

 

 

 :
 :110 °C (.5 mmHg)
 :144…150 °C
 :Ambient
 :8,III
 :220.27 g/mol
 :3261
 :GHS05
 :427-49-6
 :C₁₃H₁₆O₃
 :MFCD00019296