Order Entry
United Kingdom
ContactUsLinkComponent
2-Methyl-1-pyrroline 98%
2-Methyl-1-pyrroline 98%
Catalog # ACRO135070250
CAS Number:  
undefined
2-Methyl-1-pyrroline 98%
Catalog # ACRO135070250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4400 to 1.4420 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

About this item

 :
 :83.13 g/mol
Boiling Pt:104…105 °C (760 mmHg)
Density:0.878 g/cm³
Flash Pt:11 °C
UN:1993
ADR:3,II
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:872-32-2
Formula:C₅H₉N
MDL Number:MFCD00128806