
Ethyl-4-oxo-3-piperidinecarboxylate hydrochloride 98%
:4644-61-5
:
:C₈H₁₃NO₃
:MFCD00043267
:207.65 g/mol
:169…175 °C
:GHS07
:Ambient
- Appearance:White to yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:169 to 175 °C
- Titration with HClO4:≥97.5 %
Frequently Bought Together