
(±)-Propranolol hydrochloride 99%
:318-98-9
:3506-09-0
:C₁₆H₂₁NO₂·HCl
:MFCD00012558
:295.81 g/mol
:163…166 °C
:GHS07
:Ambient
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Melting point:163 to 166 °C
- Titration with HClO4:≥98.5 %
Frequently Bought Together