Ethyl-3-oxo-3-(pyridin-4-yl)propanoate 95%
:26377-17-3
:
:C₁₀H₁₁NO₃
:MFCD00094021
:193.20 g/mol
:53…59 °C
:Ambient
- Appearance:Off-white to yellow-brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:53 to 59 °C
- HPLC:≥94 %
Frequently Bought Together