
1-Pyrrolidinecarbonyl chloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1192-63-8
:
Formula:C₅H₈ClNO
MDL Number:MFCD00051321
Boiling Pt:75 °C (1 mmHg)
Density:1.209 g/cm³
Flash Pt:98 °C
Storage Temperature:Freezer
UN:3265
ADR:8,II
:133.575 g/mol
:GHS05
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4910 to 1.4970 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together