Order Entry
United Kingdom
ContactUsLinkComponent
 
Diacetin 50% tech. remainder triacetin and monoacetin
 
undefined
Diacetin 50% tech. remainder triacetin and monoacetin

 

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4400 to 1.4480 (20°C, 589 nm)
  • GC:
    ≥45 %

 

 

Frequently Bought Together