
1H,1H,2H,2H-Heptadecafluorodecyl acrylate 97% stabilised
Catalog # ACRO348610050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3360 to 1.3380 (20°C, 589 nm)
- GC:≥96.0 %
- Stabilizer:Approx. 15 ppm TBC and 70 ppm BHT
Specification Test Results
About this item
:
:518.17 g/mol
Boiling Pt:100 °C (4 mmHg)
Flash Pt:>110 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:27905-45-9
Formula:C₁₃H₇F₁₇O₂
MDL Number:MFCD00042306