


Dess-Martin periodinane 15% (w/w) in dichloromethane
Catalog # ACRO333110100
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- Seq:2275
Specifications
Specification Test Results
- Appearance:Colorless to light yellow Solution (May develop some turbidity or precipitate)
- Titration Iodometric:10 to 15 %
Specification Test Results
About this item
:
:424.15 g/mol
Density:1.362 g/cm³
Storage Temperature:Refrigerator
UN:1593
ADR:6.1,III
:GHS07
:GHS08
:GHS03
CAS Number:87413-09-0
Formula:C₁₃H₁₃IO₈
MDL Number:MFCD00130127