Order Entry
United Kingdom
ContactUsLinkComponent
 
 
 
 
1-Chloro-2-methylpropyl chloroformate 98%
 
undefined
 
 
 
1-Chloro-2-methylpropyl chloroformate 98%

 

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4310 to 1.4330 (20°C, 589 nm)
  • GC:
    ≥97.5 %

 

 

Frequently Bought Together