Sodium hydrosulphide hydrate, soluble, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:207683-19-0
:
Formula:H₃NaOS
MDL Number:MFCD00149796
Molecular Weight:74.073 g/mol
Melting Pt:55 °C
Flash Pt:90 °C
Storage Temperature:Ambient
UN:2949
ADR:8 II
:GHS06
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Yellow to orange to green to red to brown Deliquescent flakes (Color changes in the material may occur)
- Titration Iodimetric:≥67.0 % (NaHS)
Specification Test Results
Frequently Bought Together