Palladium 5% on alumina, dry, powder for use as a catalyst
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-05-3
:
Formula:Pd
MDL Number:MFCD03613602
Molecular Weight:106.420 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Silver grey to black Powder
- Assay:4.0 to 6.0 % (Pd)
Specification Test Results
Frequently Bought Together