
(R)-(+)-1-Amino-2-(methoxymethyl)pyrrolidine 95+%
ACRO277140010
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4635 to 1.4655 (20°C, 589 nm)
- GC:≥95.0 %
- Specific optical rotation:+76° to +84° (20°C, 589 nm) (neat)
:
:130.19 g/mol
:42 °C (1.8 mmHg)
:0.97 g/cm³
:72 °C
:GHS07
:Refrigerator
:72748-99-3
:C₆H₁₄N₂O
:MFCD00010622