Urocanic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:104-98-3
:
Formula:C₆H₆N₂O₂
MDL Number:MFCD00005203
Melting Pt:217…220 °C
:138.126 g/mol
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to beige Powder
- Infrared spectrum:Conforms
- Melting point:217 to 220 °C
- Loss on drying:≤2 %
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together