Fluor-free acids, DyLight™
Catalog # PIER62280
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 mg
- Description:DyLight™ 488 free acid
Specifications
About this item
DyLight™ fluor-free acids are non-activated forms of selected DyLight™ fluorescent dyes for use as controls and calibration factors in fluorescence imaging applications. These non-activated fluorescent dyes contain the native carboxyl group of the standard dye molecule.