Order Entry
United Kingdom
ContactUsLinkComponent
Boronic acid resin
Boronic acid resin
Catalog # PIER20244
CAS Number:  
undefined
Boronic acid resin
Catalog # PIER20244
CAS Number:  

Specifications

  • Pk:
    10 ml
  • Method / Format:
    Affinity resin
  • Prep size:
    Scalable
  • Refine target molecule:
    Ribo/oligonucleotides
  • Sample type:
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    RNA
  • Description:
    Pierce™ Boronic acid resin
  • Description:
    Pierce™ Boronic acid resin

Specifications

About this item

Pierce™ Boronic acid resin enables ribonucleotide and oligonucleotide RNA isolation.

  • Support: Beaded polyacrylamide (1800 MW exclusion limit)
  • Spacer: m-aminophenyl group
  • Loading: 100 µMol boronate per ml of settled resin
  • Capacity: at least 99% binding and recovery of 110 µMol AMP per ml of settled resin
  • Format: 50% resin slurry in 0,05% sodium azide

The immobilised boronic acid is an easy to use polyacrylamide affinity support for purification of ribonucleotides and other small molecular weight compounds that contain cis-diol groups. Boronic acid affinity chromatography has been used to isolate ribonucleoside, to purify nucleosidyl peptide, separate ribonucleosidases in tissue extracts, separate RNA and oligoribonucleotide, and determine of the amount of non-enzymatic glycosylation present in peripheral nerve from diabetic and control rats and dogs.