Bolton-Hunter reagent (sulpho-SHPP), Pierce™
Catalog # PIER27712
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
- Environmentally Preferable:
Specifications
About this item
Synonyms: 3-(4-Hydroxy-phenyl)-propionic acid-OSu, Water-Soluble Bolton-Hunter Reagent, Sulphosuccinimidyl-3 -(4-hydroxypheynyl) propionate
:
:263.249 g/mol
Storage Temperature:Freezer
CAS Number:34071-95-9
Formula:C₁₃H₁₃NO₅
MDL Number:MFCD00005515