Order Entry
United Kingdom
ContactUsLinkComponent
Alcohol dehydrogenase, MP Biomedicals
Alcohol dehydrogenase, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Alcohol dehydrogenase, MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • CAS No.:
    9031-72-5
  • Form:
    Powder
  • Enzyme name:
    Alcohol Dehydrogenase

Specifications

Frequently Bought Together