


Benzethonium chloride USP
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:121-54-0
:
Formula:C₂₇H₄₂ClNO₂
MDL Number:MFCD00011742
:448.09 g/mol
Merck Index:13,01074
Melting Pt:164…166 °C
EINECS:204-479-9
UN:3077
ADR:9,III
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Assay:97% Min.
Specification Test Results
Frequently Bought Together