Guanidinium hydrochloride ≥99%, BiotechGrade
Supplier: Spectrum Chemical
Certificates
About this item
Synonyms: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride
CAS Number:50-01-1
:
Formula:NH₂C(=NH)NH₂·HCl
MDL Number:MFCD00013026
Density:1.354 g/cm³ (20 °C)
Molecular Weight:95.53 g/mol
Melting Pt:185
Merck Index:13,04578
EINECS:200-002-3
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:99% Min.
Specification Test Results
Frequently Bought Together