Order Entry
United Kingdom
ContactUsLinkComponent
(S)-Ethyl-2-((S)-4-methyl-2,5-dioxooxazolidin-3-yl)-4-phenylbutanoate 98%
(S)-Ethyl-2-((S)-4-methyl-2,5-dioxooxazolidin-3-yl)-4-phenylbutanoate 98%
Catalog # L19355.04
CAS Number:  
(S)-Ethyl-2-((S)-4-methyl-2,5-dioxooxazolidin-3-yl)-4-phenylbutanoate 98%
Catalog # L19355.04
CAS Number:  

Specifications

  • Pk:
    2 g
  • Seq:
    3107
  • Environmentally Preferable:

Specifications

About this item

Synonyms: (S)-2-[(S)-4-Methyl-2,5-dioxooxazolidin-3-yl]-4-phenylbutanoic acid ethyl ester, Ethyl (S)-2-[(S)-4-Methyl-2,5-dioxooxazolidin-3-yl]-4-phenylbutanoate, Ethyl (S)-2-[(S)-4-Methyl-2,5-dioxo-3-oxazolidinyl]-4-phenylbutanoate, N-[1-(S)-Ethoxycarbonyl-3-phenylpropyl]-L-alanine-N-carboxyanhydride
CAS Number:84793-24-8
 :
Formula:C₁₆H₁₉NO₅
MDL Number:MFCD00272296
 :305.33 g/mol
Melting Pt:70…72 °C
EINECS:430-360-8
 :GHS05
 :GHS07
Storage Temperature:Ambient