Order Entry
United Kingdom
ContactUsLinkComponent
2-Chloro-1,1,2-trifluoroethyl difluoromethyl ether 97%
Irritant
2-Chloro-1,1,2-trifluoroethyl difluoromethyl ether 97%
Catalog # L16725.06
CAS Number:  
Irritant
2-Chloro-1,1,2-trifluoroethyl difluoromethyl ether 97%
Catalog # L16725.06
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

CAS Number:13838-16-9
 :
Formula:C₃H₂ClF₅O
MDL Number:MFCD00069095
Merck Index:14,3581
Boiling Pt:56…57 °C
Density:1.517
 :184.49 g/mol
EINECS:237-553-4
 :GHS07