Order Entry
United Kingdom
ContactUsLinkComponent
2-Amino-6-methyl-4(3H)-quinazolone 96%
Irritant
2-Amino-6-methyl-4(3H)-quinazolone 96%
Catalog # L14558.03
CAS Number:  
Irritant
2-Amino-6-methyl-4(3H)-quinazolone 96%
Catalog # L14558.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :175.19 g/mol
Melting Pt:>300 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:50440-82-9
Formula:C₉H₉N₃O
MDL Number:MFCD00236015