Filter aid, Harborlite 800
Supplier: Thermo Fisher Scientific
Jump to:
About this item
MDL No.: MFCD00197888
- MDL Number:MFCD00197888
- Density:2.3
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
MDL No.: MFCD00197888
- MDL Number:MFCD00197888
- Density:2.3
- Storage Temperature:Ambient
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



