2-Isopropyl-5-methyl-2-hexenal cis- and trans-mixture ≥96%
Catalog # L07193.18
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:50 g
- Seq:3368
Specifications
About this item
CAS Number:35158-25-9
:
Formula:C₁₀H₁₈O
MDL Number:MFCD00036616
Flash Pt:60 °C (140 °F)
Storage Temperature:Refrigerator
UN:1989
ADR:,