

Methoxycarbonylsulphenyl chloride 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:26555-40-8
:
Formula:C₂H₃ClO₂S
MDL Number:MFCD00013648
Boiling Pt:73…74 °C (100 mmHg)
Density:1.398
Flash Pt:54 °C (129 °F)
EINECS:247-801-3
ADR:8,II
:126.55 g/mol
:GHS05
:GHS02
Related Information
Frequently Bought Together