Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Dicyclopropylmethanol 97%

Dicyclopropylmethanol 97%

Dicyclopropylmethanol 97%
Certificates
Dicyclopropylmethanol 97%
Dicyclopropylmethanol 97%

2 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 14300-33-5 Molecular Formula: C7H12O Formula Weight: 112.17 Flash Point: 110°(230°F) Refractive Index: 1.4650 Boiling Point: 99-100°/12mm Beilstein Reference: 2203215 MDL No.: MFCD00019249

  • CAS Number:14300-33-5
  • Formula:C₇H₁₂O
  • MDL Number:MFCD00019249
  • Molecular Weight:112.17 g/mol
  • Boiling Pt:99…100 °C (12 mmHg)
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Ambient
  • EINECS:238-236-3
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.