AM 580
10 mg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
A retinoid derivative used for gene expression studies of leukemia-retinoic acid receptor (PHL-RAR) cell line.
- Soluble in DMSO (20mg/ml) or ethanol (10mg/ml).
White crystalline solid.
Synonyms: NSC 608001, CD336, 4-[[(5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl)carbonyl]amino]-benzoic acid, AM580, 4-(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalene-2-carboxamido)benzoic acid, RO 40-6055
- CAS Number:102121-60-8
- Formula:C₂₂H₂₅NO₃
- MDL Number:MFCD00673916
- Molecular Weight:351.44 g/mol
- Storage Temperature:Ambient
Documents
Specifications
Specifications
- Pk:10 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



