2-(3-(2,3-Dichlorophenoxy)propylamino)ethanol hydrochloride
Catalog # J64981.MA
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:300.61 g/mol
CAS Number:418788-90-6
Formula:C₁₁H₁₅Cl₂NO₂·HCl
MDL Number:MFCD06410993