Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2'-Deoxyinosine-5'-monophosphate disodium salt
2'-Deoxyinosine-5'-monophosphate disodium salt 99%
2'-Deoxyinosine-5'-monophosphate disodium salt 99%
J64271.03
:
Thermo Fisher Scientific
:
2'-Deoxyinosine-5'-monophosphate disodium salt 99%
J64271.03
:
Thermo Fisher Scientific
:
Pk:
1 g
Seq:
3047
:
:
376.17 g/mol
:
239-090-3
:
GHS07
:
14999-52-1
:
C₁₀H₁₁N₄Na₂O₇P
:
MFCD00134874