Lysis buffer, pH 8,0, for isolation of His-tag modified proteins
Catalog # J63892.AP
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:500 mL
- Description:Lysis buffer, for isolation of His-tag modified proteins
- Physical form:Liquid
- Appearance:Colourless
- pH value:8,0
Specifications
About this item
Physical Form: Liquid
Appearance: Colourless
Applications: For isolation of His-tag proteins