Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Buffers
Extraction Buffers
E. coli lysis buffer
E. coli lysis buffer
E. coli lysis buffer
J63402.AK
:
Thermo Fisher Scientific
:
E. coli lysis buffer
J63402.AK
:
Thermo Fisher Scientific
:
Pk:
250 ml
Description:
E. coli lysis buffer
Physical form:
Liquid
Appearance:
Colourless
Physical Form: Liquid
Appearance: Colourless