Destaining bags, ethidium bromide
Catalog # J62931.8U
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:25
- Description:Ethidium bromide destaining bags
Specifications
About this item
Ethidium bromide destaining bags are used to safely remove ethidium bromide from nucleotide solutions and gels during overnight treatment. Each bag extracts up to 5 mg of Ethidium Bromide from solution, thus minimising the exposure of research personnel to this material.
- The special absorbent mixture retains dye molecules in a convenient bag for removal and incineration
- Provides a safer, more economical means for disposal of hazardous waste
- Also removes Coomassie stains
Applications: For removal of ethidium bromide from nucleotide solutions and gels. Maximum capacity is 5mg ethidium bromide. Also removes Coomassie stains