Zymogram sample buffer, pH 6,6
Zymogram sample buffer, 6,6, 25 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Physical Form: Liquid
Appearance: Blue
Documents
Specifications
Specifications
- Pk:25 mL
- Description:Zymogram sample buffer
- pH value:6,6
- Cat. No.:J62774.AC
- Physical form:Liquid
- Appearance:Blue
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



