Thiostrepton ≥90%
Catalog # J62332.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
Specifications
About this item
:
:1664.887 g/mol
Merck Index:14,9364
EINECS:215-734-9
CAS Number:1393-48-2
Formula:C₇₂H₈₅N₁₉O₁₈S₅
MDL Number:MFCD00135828