RNA sample loading buffer, no ethidium bromide
Supplier: Thermo Fisher Scientific
About this item
Physical Form: Liquid
MDL No.: MFCD00213041
Applications: Formulated for electrophoresis of RNA on formaldehyde-agarose gels with or without ethidium bromide
2 Options Available Below
Product Details & Documents
Physical Form: Liquid
MDL No.: MFCD00213041
Applications: Formulated for electrophoresis of RNA on formaldehyde-agarose gels with or without ethidium bromide
Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- J61937.XF
- J61937.S0
- Physical form:
- Liquid
- Liquid
- MDL No.:
- MFCD00213041
- MFCD00213041
Specifications
Product Family Options
Product Information
- PkAvailabilityPrice
- J61937.XFRNA Sample Loading Buffer, no Ethidium BromideSpecifications:
More
Specifications
Cat. No.J61937.XFPhysical formLiquidMDL No.MFCD00213041 - J61937.S0RNA Sample Loading Buffer, no Ethidium BromideSpecifications:
More
Specifications
Cat. No.J61937.S0Physical formLiquidMDL No.MFCD00213041
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



