Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Oligomycin

Oligomycin

10 mg
Catalog # J61898.MA
Irritant
Oligomycin
Catalog # J61898.MA
CAS Number:  1404-19-9
Oligomycin
Oligomycin

10 mg

CAS Number:  1404-19-9
Price
£203.00
Log in for availability
1 * 10 mg
Product Details & Documents

CAS No.: 1404-19-9 Molecular Formula: C45H74O11 Formula Weight: 791.07 Physical Form: Powder Appearance: White Solubility: Sparingly soluble in water. Soluble in DMSO (20mg/ml) and ethanol (30mg/ml) Beilstein Reference: 7615308 MDL No.: MFCD01779388

Applications: Inhibits respiration in mitochondria. Also inhibits ATP synthase

Synonyms:  Oligomycin A, B, C, Oligomycin complex
  • CAS Number:1404-19-9
  • Formula:C₄₅H₇₄O₁₁
  • MDL Number:MFCD01779388
  • Molecular Weight:789.3 g/mol
  • Merck Index:14,6833
  • Storage Temperature:Freezer
  • EINECS:215-767-9
Specifications

Specifications

  • Pk:
    10 mg

Specifications