8-Cyclopentyl-1,3-dimethylxanthine
Catalog # J61565.MC
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:100 mg
Specifications
About this item
:
:248.286 g/mol
Melting Pt:250…252 °C
CAS Number:35873-49-5
Formula:C₁₂H₁₆N₄O₂
MDL Number:MFCD00055116