Order Entry
United Kingdom
ContactUsLinkComponent
NP-40 lysis buffer, high salt
NP-40 lysis buffer, high salt
Catalog # J61428.AK
CAS Number:  
NP-40 lysis buffer, high salt
Catalog # J61428.AK
CAS Number:  

Specifications

  • Pk:
    250 ml
  • Description:
    NP-40 lysis buffer, high salt
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Viscous liquid
  • Appearance:
    Colourless

Specifications

About this item

Storage Temperature: 2 to 8 °C
Physical Form: Viscous liquid
Appearance: Colourless