Order Entry
United Kingdom
ContactUsLinkComponent
Doxofylline
Irritant
Doxofylline
Catalog # J60575.06
CAS Number:  
Irritant
Doxofylline
Catalog # J60575.06
CAS Number:  

Specifications

  • Pk:
    5 g

Specifications

About this item

CAS Number:69975-86-6
 :
Formula:C₁₁H₁₄N₄O₄
MDL Number:MFCD00865218
Merck Index:14,3438
Melting Pt:144…146 °C
 :266.26 g/mol
EINECS:274-239-6
 :GHS07
Storage Temperature:Ambient