Horseradish peroxidase
Supplier: Thermo Fisher Scientific
Certificates
About this item
Specifications
- CAS No.:9003-99-0
- Species:Horseradish
- Storage conditions:–30...–10 °C
- Form:Off-White to pale brown lyophilized powder
- MDL No.:MFCD00071339
- Enzyme name:Peroxidase
- Solubility:Soluble in water or dilute buffer
Specifications
Frequently Bought Together